Learn More
CPC Scientific Z-Leu-Leu-Leu-CHO 5MG
SDP
Supplier:  CPC Scientific CALI001A
Encompass
A potent and reversible cell permeable proteasome inhibitor (Kj 4 nM) of 20S proteasome and calpain. Also shown to initiate neurite outgrouth in PC12 cells with an optimal concentration set at 20 nM.SEQUENCE: Z-Leu-Leu-Leu-CHOONE-LETTER SEQUENCE: Z-LLL-CHOMOLECULAR FORMULA: C26H41N3O5MOLECULAR WEIGHT: 475.63STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [133407-82-6]SYNONYMS: Calpain Inhibitor IV, MG 132, MG132RESEARCH AREA: CancerREFERENCES:D.H. Lee et al., J. Biol. Chem., 271, 27280 (1996)K. L. Rock et al, Cell, 78, 761 (1994)SKU(s): CALI-001A
Catalog No. 50-210-8863