Learn More
TARGETMOL CHEMICALS INC MG 149 10MG
SDP
Catalog No. 502579330
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MG 149 (Tip60 HAT inhibitor) is a selective and potent Tip60 inhibitor with IC50 of 74 uM similar potentcy for MOF(IC50= 47 uM). purity: 99%
Catalog No. 50-257-9330 Supplier TARGETMOL CHEMICALS INC Supplier No. T658410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MG 149 (Tip60 HAT inhibitor) is a selective and potent Tip60 inhibitor with IC50 of 74 uM similar potentcy for MOF(IC50= 47 uM). purity: 99%