Learn More
Medchemexpress LLC MG-2119 | 1065500-40-4 | 99.7% | 487.55 | 100 MG
SDP
Catalog No. 5000349687
Encompass_Preferred
MG-2119 is a potent monomeric tau and α-syn aggregation inhibitor. It is a potential agent for neurological disorders research.
  • Potent monomeric tau and α-syn aggregation inhibitor
  • Potential agent for neurological disorders research
  • Useful in neurological, eye, or ear disease research
  • Relevant for neurodegenerative disease and neuronal signaling studies
Catalog No. 50-003-49687 Supplier Medchemexpress LLC Supplier No. HY139307100MG
MG-2119 is a potent monomeric tau and α-syn aggregation inhibitor. It is a potential agent for neurological disorders research.
  • Potent monomeric tau and α-syn aggregation inhibitor
  • Potential agent for neurological disorders research
  • Useful in neurological, eye, or ear disease research
  • Relevant for neurodegenerative disease and neuronal signaling studies