Learn More
Strem, An Ascensus Company CAS# [NO CAS#]. 5g. Bis(N,N'-di-sec-butylacetamidinato)magnesium, 99%.
SDP
Supplier:  Strem, An Ascensus Company 1208455G
Encompass
5g. Bis(N,N'-di-sec-butylacetamidinato)magnesium, 99%. . Molecular Weight: 362.88. Molecular Formula: C20H42MgN4. Color/form: colorless to pale yellow liq. Strem# 12-0845. www.strem.com/catalog/v3/12-0845/
Catalog No. 50-197-9354