Learn More
Andwin Scientific MG132 1MG
SDP
Catalog No. NC9937881
Encompass_Preferred
MG-132 is a chemical that performs as a cell-permeable, potent, reversible proteasome inhibitor. This product is a white solid chemical that is in powder form. It has a Health Hazard NFPA Rating of 0, a Fire Hazard NFPA Rating of 1, and a Reactivity NFPA Rating of 0. This is packaged and sold in a 1 milligram glass bottle.
Catalog No. NC9937881 Supplier Andwin Scientific Supplier No. 474790
MG-132 is a chemical that performs as a cell-permeable, potent, reversible proteasome inhibitor. This product is a white solid chemical that is in powder form. It has a Health Hazard NFPA Rating of 0, a Fire Hazard NFPA Rating of 1, and a Reactivity NFPA Rating of 0. This is packaged and sold in a 1 milligram glass bottle.