Learn More
Eurofins Discoverx MG132, 5 mg. (92-1098)
Supplier:  Eurofins Discoverx 921098
Encompass
MG132 is a synthetic inhibitor that potently, selectively, and reversibly inhibits the proteasome (IC50=100 nM), assisting in ROS formation and apoptosis. Also activates JNK1, and inhibits calpain (IC50=1.2 µM). Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9635