Learn More
INTACT GENOMICS INC MG1655 Chemically Competent Cells 12x200µl
Catalog No. 5000083046
Encompass
Intact Genomics (ig) MG1655 Chemically Competent Cells are suitable for routine DNA transformation and other research purposes. MG1655 is the “wild type” K-12 strain with minimal genetic manipulation having only been cured of the temperate bacteriophage lambda and F plasmid by ultraviolet light and acridine orange (1).ig MG1655 chemically competent cells have the following genomic features: • Cured of the temperate bacteriophage lambda and F plasmid • A frameshift at the end of rph • ilvG- mutation is a frameshift that knocks out acetohydroxy acid synthase II • rfb-50 mutation is an IS5 insertion that results in the absence of O-antigen synthesis
Catalog No. 50-000-83046 Supplier INTACT GENOMICS INC Supplier No. 107648
Intact Genomics (ig) MG1655 Chemically Competent Cells are suitable for routine DNA transformation and other research purposes. MG1655 is the “wild type” K-12 strain with minimal genetic manipulation having only been cured of the temperate bacteriophage lambda and F plasmid by ultraviolet light and acridine orange (1).ig MG1655 chemically competent cells have the following genomic features: • Cured of the temperate bacteriophage lambda and F plasmid • A frameshift at the end of rph • ilvG- mutation is a frameshift that knocks out acetohydroxy acid synthase II • rfb-50 mutation is an IS5 insertion that results in the absence of O-antigen synthesis