Learn More
Enzo Life Sciences Mg2+/ATP Activating Solution (100 µl)
SDP
Catalog No. NC1275801
Encompass_Preferred
This solution contains pre-coupled Mg-ATP for immediate use as an energy source for a range of ATP-dependent reactions (e.g. ubiquitin thioester conjugation, proteasomal degradation, SUMO conjugation). Formulation: Liquid. In 50mM TRIS-Cl, pH 7.5, containing 0.1M ATP and 0.1M MgCl2. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C
Catalog No. NC1275801 Supplier Enzo Life Sciences Supplier No. BMLEW98050100
This solution contains pre-coupled Mg-ATP for immediate use as an energy source for a range of ATP-dependent reactions (e.g. ubiquitin thioester conjugation, proteasomal degradation, SUMO conjugation). Formulation: Liquid. In 50mM TRIS-Cl, pH 7.5, containing 0.1M ATP and 0.1M MgCl2. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C