Learn More
MOLTOX Molecular Toxicology Inc Minimal Glucose Agar, 30 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948614
Encompass
Our Minimal Glucose Agars are specifically formulated for use in bacterial mutagenicity assays. Along with the standard formulation as described by Maron & Ames, Mutat. Res. 113:173, 1983, MOLTOX offers various other formulations such as histidine and biotin supplemented bottom agar. Our MGA""s are offered in case quantities or individual sleeves. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.
Catalog No. 50-948-614 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 21400.2
Our Minimal Glucose Agars are specifically formulated for use in bacterial mutagenicity assays. Along with the standard formulation as described by Maron & Ames, Mutat. Res. 113:173, 1983, MOLTOX offers various other formulations such as histidine and biotin supplemented bottom agar. Our MGA""s are offered in case quantities or individual sleeves. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.