Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MGC DAU Control Sets

Catalog No. 23278255 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Clinical DAU Control Set
Optional DAU Control Set
Primary DAU Control Set
Select DAU Control Set
Specialty DAU Control Set
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-278-255 Specialty DAU Control Set
23-278-254 Primary DAU Control Set
23-278-253 Clinical DAU Control Set
23-278-249 Select DAU Control Set
23-278-247 Optional DAU Control Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-278-255 Supplier Thermo Scientific™ Supplier No. 1815440
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets.

Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets. These controls contain high and low level controls at drug concentrations 25% below and above screening assay cutoffs to ensure quality laboratory results.

Key Features
  • Multi-constituent
  • For use in the validation of drug of abuse assays in human urine
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Amphetamine, Secobarbital, Nitrazepam, Benzoylecgonine, Benzoylecgonine, d-Methamphetamine, EDDP, Methadone Metabolite (EDDP), Morphine, Nitrazepam, Opiate 300 (Morphine), Secobarbital
Sample Type Urine
Type Specialty DAU Control Set
Includes Amphetamine: 375ng/mL and 625ng/mL; Barbiturate: 150ng/mL and 250ng/mL; Benzodiazepine: 150ng/mL and 250ng/mL; Cocaine: 112.5ng/mL and 187.5ng/mL; Methadone Metabolite: 75ng/mL and 125ng/mL; and Opiate 300: 225ng/mL and 375ng/mL
Quantity 3 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.