Learn More
Aviva Systems Biology Corp MGC20983 Antibody - middle region (ARP52787_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC20983. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGC20983. Alternate names: FLJ31801, MGC20983ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60831