Learn More
Aviva Systems Biology Corp MGC27016 Antibody - N-terminal region (ARP41134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC27016. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MGC27016. Alternate names: CT68, MGC27016ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53279