Learn More
Aviva Systems Biology Corp MGC29891 Antibody - C-terminal region (ARP50783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC29891. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MGC29891. Alternate names: RP11-68I18.1, MGC29891ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60433