Learn More
Aviva Systems Biology Corp MGC33407 Antibody - middle region (ARP55773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC33407. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGC33407. Alternate names: HSD21, MGC33407ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61605