Learn More
Aviva Systems Biology Corp MGC33926 Antibody - middle region (ARP42935_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42935_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC33926. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGC33926. Alternate names: TMEM178, MGC33926ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58367