Learn More
Aviva Systems Biology Corp MGC39633 Antibody - N-terminal region (ARP44931_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44931_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC39633. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MGC39633. Alternate names: MBC1, MGC39633ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58940