Learn More
Aviva Systems Biology Corp MGC4172 Antibody - C-terminal region (ARP49813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC4172. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MGC4172. Alternate names: ARPG836, FLJ39232, SDR24C1, MGC4172ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60194