Learn More
Aviva Systems Biology Corp MGC42174 Antibody - N-terminal region (ARP41145_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41145_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC42174. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MGC42174. Alternate names: FAM6A, MGC42174ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57737