Learn More
Aviva Systems Biology Corp MGC45491 Antibody - middle region (ARP55574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC45491. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGC45491. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61542