Learn More
Aviva Systems Biology Corp MGC4618 Antibody - N-terminal region (ARP34360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC4618. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MGC4618. Alternate names: ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55354