Learn More
Aviva Systems Biology Corp MGC46336 Antibody - N-terminal region (ARP40116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC46336. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MGC46336. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57296