Learn More
Aviva Systems Biology Corp CCDC187 Antibody - N-terminal region (ARP55951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC50722. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MGC50722. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rat
Catalog No. 50-014-61673