Learn More
Aviva Systems Biology Corp MGC51025 Antibody - C-terminal region (ARP55784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC51025. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MGC51025. Alternate names: MGC51025ApplicationWB. Species reactivity: Human
Catalog No. 50-286-080