Learn More
Aviva Systems Biology Corp MGC87631 Antibody - middle region (ARP54385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC87631. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGC87631. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-285-918