Learn More
Apexbio Technology LLC MGCD-265 875337-44-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093779
Encompass_Preferred
MGCD-265 (CAS 875337-44-3) is a multi-targeted ATP-competitive small molecule inhibitor primarily targeting c-Met and the vascular endothelial growth factor receptors VEGFR1 VEGFR2 and VEGFR3 with IC50 values of 1 nM 3 nM 3 nM and 4 nM respectively In preclinical non-small cell lung cancer (NSCLC) xenograft models including models with TKI-resistant EGFR T790M mutations MGCD-265 in combination with agents such as paclitaxel docetaxel or erlotinib demonstrated enhanced antitumor activity and antiangiogenic effects compared to monotherapy MGCD-265 has been investigated in phase I clinical trials as a single agent and in combination with docetaxel or erlotinib in patients with advanced solid tumors
Catalog No. 50-000-93779 Supplier Apexbio Technology LLC Supplier No. A50575.1
MGCD-265 (CAS 875337-44-3) is a multi-targeted ATP-competitive small molecule inhibitor primarily targeting c-Met and the vascular endothelial growth factor receptors VEGFR1 VEGFR2 and VEGFR3 with IC50 values of 1 nM 3 nM 3 nM and 4 nM respectively In preclinical non-small cell lung cancer (NSCLC) xenograft models including models with TKI-resistant EGFR T790M mutations MGCD-265 in combination with agents such as paclitaxel docetaxel or erlotinib demonstrated enhanced antitumor activity and antiangiogenic effects compared to monotherapy MGCD-265 has been investigated in phase I clinical trials as a single agent and in combination with docetaxel or erlotinib in patients with advanced solid tumors