Learn More
Medchemexpress LLC MGCD-265 analog | 875337-44-3 | 98.57% | 517.60 | 1 ML
SDP
Catalog No. 5000346550
Encompass_Preferred
MGCD-265 analog is a potent and oral active inhibitor of c-Met and VEGFR2 tyrosine kinases, with IC50s of 29 nM and 10 nM, respectively, and has significant antitumor activity.
  • Potent inhibitor of c-Met and VEGFR2 tyrosine kinases.
  • Oral active.
  • Exhibits significant antitumor activity.
  • Inhibits A549 cells migration and DU145 cells scattering.
  • Inhibits HUVEC ERK phosphorylation and HUVEC proliferation.
Catalog No. 50-003-46550 Supplier Medchemexpress LLC Supplier No. HY1099110MM/1ML
MGCD-265 analog is a potent and oral active inhibitor of c-Met and VEGFR2 tyrosine kinases, with IC50s of 29 nM and 10 nM, respectively, and has significant antitumor activity.
  • Potent inhibitor of c-Met and VEGFR2 tyrosine kinases.
  • Oral active.
  • Exhibits significant antitumor activity.
  • Inhibits A549 cells migration and DU145 cells scattering.
  • Inhibits HUVEC ERK phosphorylation and HUVEC proliferation.