Learn More
Selleck Chemical LLC MGCD-265 analog 50mg 875337-44-3
SDP
Supplier:  Selleck Chemical LLC S136150MG
Encompass_Preferred
MGCD-265 is a potent, multi-target and ATP-competitive inhibitor of c-Met and VEGFR1/2/3 with IC50 of 1 nM, 3 nM/3 nM/4 nM, respectively; also inhibits Ron and Tie2. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-676-0