Learn More
Lonza Walkersville MgCl2 10 mM Solution
Catalog No. NC9556098
Encompass
MgCl2 can be used as the sample diluent when attempting to overcome inhibitory chelation effect. Examples of samples which chelate divalent cations include heparin and EDTA. MgCl2 may be used to prepare samples for any LAL assay. Contains <0.005 EU/ml endotoxin.Content:30 mL/vial
Catalog No. NC9556098 Supplier Lonza Walkersville Supplier No. S50641
MgCl2 can be used as the sample diluent when attempting to overcome inhibitory chelation effect. Examples of samples which chelate divalent cations include heparin and EDTA. MgCl2 may be used to prepare samples for any LAL assay. Contains <0.005 EU/ml endotoxin.Content:30 mL/vial