Learn More
AG Scientific Inc MgCl2 Hexahydrate, 1 KG
Catalog No. 502236858
Encompass_Preferred
Magnesium Chloride Hexahydrate is a widely used reagent in chemistry and molecular biology as a source of magnesium ions. Not for human therapeutic use or for medicinal purposes. For research applications only.CAS Number: 7791-18-6Molecular Weight: 203.3 DaChemical Formula: Cl2Mg 6H2OSolubility: Water (167 g in 100 mL water)Storage Temperature: Room Temperature
Catalog No. 50-223-6858 Supplier AG Scientific Inc Supplier No. M26811KG
Magnesium Chloride Hexahydrate is a widely used reagent in chemistry and molecular biology as a source of magnesium ions. Not for human therapeutic use or for medicinal purposes. For research applications only.CAS Number: 7791-18-6Molecular Weight: 203.3 DaChemical Formula: Cl2Mg 6H2OSolubility: Water (167 g in 100 mL water)Storage Temperature: Room Temperature