Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFG Tray Dolly for Nesting Storage Boxes

Catalog No. 1523995 Shop All Molded Fiber Glass Tray Company Products
Change view
Click to view available options
For Use With (Equipment):
Nesting Storage Boxes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
15-239-95 Nesting Storage Boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-239-95 Supplier MFG Tray Supplier No. 87014851361075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dolly for 870, 875, 880 stacking boxes

  • Impervious to radiation, cutting oils, greases, solvents, detergents, mild acids and alkaline solutions within a pH rating of 3.0 to 10.0
  • Temperature rating of -51° to +121°C (-60° to +250°F)
  • Smooth outside surface and radial edges
  • Tough composite construction
  • Nesting design saves space
  • Machine integration and robotic applications

Specifications

Type Dolly
Color Gray
For Use With (Application) Nesting Storage Boxes
For Use With (Equipment) Nesting Storage Boxes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.