Learn More
Cayman Chemical Miglitol-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502990089
Encompass_Preferred
An internal standard for the quantification of miglitol by GC- or LC-MS
Catalog No. 50-299-0089 Supplier Cayman Chemical Supplier No. 301391 MG
An internal standard for the quantification of miglitol by GC- or LC-MS