Learn More
Medchemexpress LLC mGluR3 modulator-1 | 374548-18-2 | MFCD01957075 | 98.7% | 271.36 g·mol⁻1 | C16H21N3O | 10 MG
SDP
Catalog No. 5000286540
Encompass_Preferred
mGluR3 modulator-1 is a small-molecule modulator of the metabotropic glutamate receptor 3 (mGluR3) intended for research use. It demonstrates activity in cellular calcium mobilization assays (EC50 = 1-10 μM) and is supplied with a reported high purity and defined molecular properties suitable for in vitro pharmacology and receptor profiling studies.
  • Modulates mGluR3 receptor activity in cell-based assays.
  • Demonstrated EC50 of 1-10 μM in HEK293T-mGluR-Gqi5 calcium mobilization assay.
  • High purity suitable for research applications.
  • Available in small mass packages for screening and assay work.
  • Includes chemical identifiers for traceability, such as CAS 374548-18-2.
Catalog No. 50-002-86540 Supplier Medchemexpress LLC Supplier No. HY15313210MG
mGluR3 modulator-1 is a small-molecule modulator of the metabotropic glutamate receptor 3 (mGluR3) intended for research use. It demonstrates activity in cellular calcium mobilization assays (EC50 = 1-10 μM) and is supplied with a reported high purity and defined molecular properties suitable for in vitro pharmacology and receptor profiling studies.
  • Modulates mGluR3 receptor activity in cell-based assays.
  • Demonstrated EC50 of 1-10 μM in HEK293T-mGluR-Gqi5 calcium mobilization assay.
  • High purity suitable for research applications.
  • Available in small mass packages for screening and assay work.
  • Includes chemical identifiers for traceability, such as CAS 374548-18-2.