Learn More
Bachem H-Met-Gly-Pro-AMC · HCl
SDP
Supplier:  Bachem 4033312.0100
Encompass_Preferred
H-Met-Gly-Pro-AMC · HCl, 1926163-53-2, C22H28N4O5S · HCl, Mr 497.02, 100mg. MGP-AMC, fluorogenic substrate for methionine aminopeptidases 1D and 2.
Catalog No. 50-194-7057