Learn More
AnaSpec Mgp100 (25-33), mouse, 1mg
Supplier:  AnaSpec AS64752
Encompass
Mgp100 (25-33), mouse[EGSRNQDWL] ; MW 1104.2; (1 mg) This peptide sequence is found in residues 25 to 33 of the mouse self/tumor antigen glycoprotein (mgp100). This fragment is a H-2Dbrestricted epitope recognized by CD8+ T cells. Mgp100 is an enzyme normally involved in pigment synthesis, and the epitope fragment is typically expressed in both normal melanocytes and melanoma cells.
Catalog No. NC0888172