Learn More
Aviva Systems Biology Corp MGST1 Antibody - N-terminal region (ARP60340_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60340_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGST1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MGST1. Alternate names: GST12, MGC14525, MGST, MGST-IApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-011