Learn More
Aviva Systems Biology Corp MGST3 Antibody - N-terminal region (ARP60342_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60342_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGST3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MGST3. Alternate names: GST-IIIApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-012