Learn More
Rheosense Inc MGVS60 CALIBRATION FLUID 100ML
SDP
Catalog No. NC1537000
Encompass
MGVS60 Water based calibration fluid (6 cP) - 100 mL
Catalog No. NC1537000 Supplier Rheosense Inc Supplier No. H3005
MGVS60 Water based calibration fluid (6 cP) - 100 mL