Learn More
STEMCELL Technologies Anti-Mouse MHC Class II (I-A/I-E), Clone M5/114.15.2, PE, Size: 25 ug
SDP
Supplier:  STEMCELL Technologies 1000292
Encompass
Rat monoclonal IgG2b antibody against mouse MHC Class II (I-A/I-E) PE-conjugated
Catalog No. 50-239-0150