Learn More
Enzo Life Sciences MHC Class I monoclonal antibody (W6/32) (R-PE conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805711RC100
Encompass_Preferred
Clone: W6/32. Isotype: IgG2a. Immunogen: Membrane of human tonsil cells. Species reactivity: Human, Monkey, Cat, Bovine. Specificity: Recognizes virtually all nucleated human cells. Suitable as positive control for HLA tissue typing and crossmatching. Applications: Flow Cytometry. Purity: Protein G-affinity purified. The purified antibody is conjugated with R-Phycoerythrin (PE). The conjugate is purified by size-exclusion chromatography. Formulation: Liquid. In PBS containing 15mM sodium azide and 0.2% BSA.
Catalog No. 50-201-0066