Learn More
Enterprise Technology Solutions MHMS Biohazard Waste Bag,30-33 Gallon,31"x43",50/BX,Red
SDP
Catalog No. 5000790337
Encompass_Preferred
Bags labeled "Infectious Waste" provide for immediate, safe disposal of infectious waste. Each 1.5 mil thick bag displays a large biohazard symbol and warning message in English and Spanish. Bags also meet OSHA requirements.
Catalog No. 50-007-90337 Supplier Enterprise Technology Solutions Supplier No. MHMRIWB142143
Bags labeled "Infectious Waste" provide for immediate, safe disposal of infectious waste. Each 1.5 mil thick bag displays a large biohazard symbol and warning message in English and Spanish. Bags also meet OSHA requirements.