Learn More
ATCC MH-S; Alveolar Macrophage; Mouse (Mus musculus)
Catalog No. 502384702
Encompass
MH-S is a macrophage, alveolar cell line that was isolated from the lung of a 7-week-old, male mouse. This cell line was deposited by I Mbawuike, HB Herscowitz, and can be used in immunology research.
Catalog No. 50-238-4702 Supplier ATCC Supplier No. CRL2019
MH-S is a macrophage, alveolar cell line that was isolated from the lung of a 7-week-old, male mouse. This cell line was deposited by I Mbawuike, HB Herscowitz, and can be used in immunology research.