Learn More
Selleck Chemical LLC MHY-1685-25mg
SDP
Supplier:  Selleck Chemical LLC S761425MG
Encompass_Preferred
MHY-1685 is a novel mTOR inhibitor, which attenuates senescence by modulating autophagy through the inhibition of mTOR in human cardiac stem cells.
Catalog No. 50-238-9835