Learn More
Selleck Chemical LLC MHY1485 10mg 326914-06-1
SDP
Supplier:  Selleck Chemical LLC S781110MG
Encompass_Preferred
MHY1485 is a potent, and cell-permeable mTOR activator, and also potently inhibits autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5285