Learn More
Medchemexpress LLC HY-B0795 10mg , MHY1485 CAS:326914-06-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB079510MG
Encompass_Preferred
HY-B0795 10mg MHY1485 CAS: 326914-06-1 MHY1485 is a potent cell-permeable mTOR activator that targets the ATP domain of mTOR. MHY1485 inhibits autophagy by suppression of fusion between autophagosomes and lysosomes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9817