Learn More
Apexbio Technology LLC MHY1485 326914-06-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063934
Encompass_Preferred
MHY1485 (CAS 326914-06-1) is a potent activator of mammalian target of rapamycin (mTOR) a serine/threonine kinase critical for cellular growth metabolism and survival pathways By activating mTOR signaling MHY1485 inhibits autophagy initiation and disrupts the fusion step between autophagosomes and lysosomes resulting in accumulation of autophagic marker LC3-II and enlarged autophagic structures In cultured rat Ac2F hepatic cells MHY1485 suppresses basal and starvation-induced autophagic flux Additionally MHY1485 treatment promotes ovarian follicular development and increases explant growth in mouse ovarian culture models providing a useful tool in biomedical research of mTOR signaling and reproductive biology
Catalog No. 50-000-63934 Supplier Apexbio Technology LLC Supplier No. B58535.1
MHY1485 (CAS 326914-06-1) is a potent activator of mammalian target of rapamycin (mTOR) a serine/threonine kinase critical for cellular growth metabolism and survival pathways By activating mTOR signaling MHY1485 inhibits autophagy initiation and disrupts the fusion step between autophagosomes and lysosomes resulting in accumulation of autophagic marker LC3-II and enlarged autophagic structures In cultured rat Ac2F hepatic cells MHY1485 suppresses basal and starvation-induced autophagic flux Additionally MHY1485 treatment promotes ovarian follicular development and increases explant growth in mouse ovarian culture models providing a useful tool in biomedical research of mTOR signaling and reproductive biology