Learn More
Medchemexpress LLC MI-1061 TFA | 1410737-35-7 | 97.5% | 696.47 | C32H27Cl2F4N3O6 | 10MM 1ML
SDP
Catalog No. 5000235720
Encompass_Preferred
MI-1061 TFA is the trifluoroacetate salt of MI-1061, a potent, orally bioavailable inhibitor of the MDM2-p53 interaction (reported IC50 = 4.4 nM; Ki = 0.16 nM). The compound is offered as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro assays, and the manufacturer provides solubility and in vivo formulation guidance. Typical uses include biochemical and cell-based studies of p53 pathway modulation.
  • Potent MDM2-p53 interaction inhibitor (IC50 4.4 nM).
  • Available as solid and as a 10 mM solution in DMSO for immediate use.
  • High purity (approximately 97.5%), suitable for research assays.
  • Soluble in DMSO (reported solubility 120 mg/mL), enabling concentrated stocks.
  • Manufacturer provides recommended stock concentrations and in vivo vehicle guidance.
Catalog No. 50-002-35720 Supplier Medchemexpress LLC Supplier No. HY125858A10MM 1ML
MI-1061 TFA is the trifluoroacetate salt of MI-1061, a potent, orally bioavailable inhibitor of the MDM2-p53 interaction (reported IC50 = 4.4 nM; Ki = 0.16 nM). The compound is offered as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro assays, and the manufacturer provides solubility and in vivo formulation guidance. Typical uses include biochemical and cell-based studies of p53 pathway modulation.
  • Potent MDM2-p53 interaction inhibitor (IC50 4.4 nM).
  • Available as solid and as a 10 mM solution in DMSO for immediate use.
  • High purity (approximately 97.5%), suitable for research assays.
  • Soluble in DMSO (reported solubility 120 mg/mL), enabling concentrated stocks.
  • Manufacturer provides recommended stock concentrations and in vivo vehicle guidance.