Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mi-1061 Tfa | 1410737-35-7 | 97.5% | 696.47 | C32H27Cl2F4N3O6 | 25MG
SDP

Catalog No. 5000235962
Encompass_Preferred

MI-1061 TFA is the trifluoroacetate (TFA) salt of MI-1061, a potent, orally bioavailable inhibitor of the MDM2-p53 protein-protein interaction. It activates p53, induces apoptosis in tumor models, and shows selective activity in p53 wild-type cell lines.

  • Potent biochemical MDM2-p53 interaction inhibitor (IC50 = 4.4 nM; Ki = 0.16 nM).
  • Selective cellular activity with IC50 = 100 nM in SJSA-1 and 250 nM in HCT-116 p53+/+; >10,000 nM in p53-/- cells.
  • Trifluoroacetate salt presented as a solid form.
  • Molecular weight 696.47 and molecular formula C32H27Cl2F4N3O6.
  • Intended for research use as a tool compound to study p53 activation and antitumor effects.
  • Purity listed as 97.47%.

Catalog No. 50-002-35962 Supplier Medchemexpress LLC Supplier No. HY125858A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MI-1061 TFA is the trifluoroacetate (TFA) salt of MI-1061, a potent, orally bioavailable inhibitor of the MDM2-p53 protein-protein interaction. It activates p53, induces apoptosis in tumor models, and shows selective activity in p53 wild-type cell lines.

  • Potent biochemical MDM2-p53 interaction inhibitor (IC50 = 4.4 nM; Ki = 0.16 nM).
  • Selective cellular activity with IC50 = 100 nM in SJSA-1 and 250 nM in HCT-116 p53+/+; >10,000 nM in p53-/- cells.
  • Trifluoroacetate salt presented as a solid form.
  • Molecular weight 696.47 and molecular formula C32H27Cl2F4N3O6.
  • Intended for research use as a tool compound to study p53 activation and antitumor effects.
  • Purity listed as 97.47%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.