Learn More
Medchemexpress LLC HY-19319 5mg Medchemexpress, MI-136 CAS:1628316-74-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY193195MG
Encompass_Preferred
Medchemexpress, HY-19319 5mg MI-136 CAS:1628316-74-4 MI-136 is an inhibitor of the menin-MLL protein-protein interaction (PPI), with an IC50 of 31 nM and a Kd of 23.6 nM. MI-136 shows to block AR signaling and has the potential for the study in castration-resistant tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2115