Learn More
Apexbio Technology LLC MI-2(Synonyms: Menin-MLL Inhibitor MI-2, MI2, MI 2, Menin inhibitor MI-2), 200mg, CAS: 1271738-62-5.
SDP
Catalog No. 5000060311
Encompass_Preferred
MI-2 (CAS 1271738-62-5) is a small molecule inhibitor targeting the menin-MLL protein-protein interaction with an IC50 of 0 45 M The formation of menin-MLL fusion protein complexes resulting from chromosomal rearrangements plays an essential role in the pathogenesis of acute myeloid leukemia (AML) and acute lymphoblastic leukemia (ALL) MI-2 selectively binds wild-type menin disrupting its interaction with MLL-fusion proteins such as MLL-AF9 thereby inhibiting cellular proliferation and colony formation in MLL-AF9-transduced mouse bone marrow cells MI-2 serves as a research tool in studies of leukemia biology and menin-associated oncogenesis
Catalog No. 50-000-60311 Supplier Apexbio Technology LLC Supplier No. A3602200
MI-2 (CAS 1271738-62-5) is a small molecule inhibitor targeting the menin-MLL protein-protein interaction with an IC50 of 0 45 M The formation of menin-MLL fusion protein complexes resulting from chromosomal rearrangements plays an essential role in the pathogenesis of acute myeloid leukemia (AML) and acute lymphoblastic leukemia (ALL) MI-2 selectively binds wild-type menin disrupting its interaction with MLL-fusion proteins such as MLL-AF9 thereby inhibiting cellular proliferation and colony formation in MLL-AF9-transduced mouse bone marrow cells MI-2 serves as a research tool in studies of leukemia biology and menin-associated oncogenesis