Learn More
Selleck Chemical LLC MI-2
SDP
Catalog No. 5000004505
Encompass_Preferred
MI-2 (MALT1 inhibitor) is an irreversible MALT1 inhibitor with IC50 of 5 84 M
Catalog No. 50-000-04505 Supplier Selleck Chemical LLC Supplier No. S742950MG
MI-2 (MALT1 inhibitor) is an irreversible MALT1 inhibitor with IC50 of 5 84 M